Assay Details
Target Gene Details
Entrez Gene ID: | 8992 |
Gene Name: | ATPase H+ transporting V0 subunit e1 |
Gene Aliases: |
ATP6H, ATP6V0E, M9.2, Vma21, Vma21p |
Location: |
Chr.5:172983760-173034897 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V0E1 | NM_003945.3 | NP_003936.1 | ||
AK312068.1 | ||||
BC119714.1 | ||||
BC119715.1 | ||||
BF691381.1 | ||||
CR456856.1 | CAG33137.1 | |||
CR542131.1 | CAG46928.1 | |||
Y15286.1 | CAA75571.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830555 | Chr.5:172866675 - 173034104 on Build GRCh38 | Loss | ATP6V0E1 LOC100268168 ERGIC1 SNORA74B RPL26L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |