Assay Details
Target Gene Details
Entrez Gene ID: | 100861519 |
Gene Name: | GDNF antisense RNA 1 (head to head) |
Gene Aliases: |
GDNFOS |
Location: |
Chr.5:37840028-37875799 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GDNF-AS1 | NR_103441.1 | |||
XM_017008916.1 | XP_016864405.1 | |||
JF824130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1593e212 | Chr.5:37832461 - 37848988 on Build GRCh38 | Loss | GDNF-AS1 GDNF |
esv3604748 | Chr.5:37720912 - 37848930 on Build GRCh38 | Gain | WDR70 GDNF-AS1 GDNF |