Assay Details
Target Gene Details
Entrez Gene ID: | 6502 |
Gene Name: | S-phase kinase-associated protein 2, E3 ubiquitin protein ligase |
Gene Aliases: |
FBL1, FBXL1, FLB1, p45 |
Location: |
Chr.5:36151989-36192792 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SKP2 | NM_001243120.1 | NP_001230049.1 | ||
NM_005983.3 | NP_005974.2 | |||
NM_032637.3 | NP_116026.1 | |||
XM_011514081.2 | XP_011512383.1 | |||
XM_011514082.2 | XP_011512384.1 | |||
XM_011514083.2 | XP_011512385.1 | |||
XM_017009753.1 | XP_016865242.1 | |||
AB050979.1 | BAB87200.1 | |||
AB050980.1 | BAB87201.1 | |||
AB050981.1 | BAB87202.1 | |||
AK291255.1 | ||||
AK295152.1 | ||||
AK296223.1 | ||||
AY029177.1 | AAK31593.1 | |||
BC001441.2 | AAH01441.1 | |||
BC007441.2 | AAH07441.1 | |||
U33761.1 | AAC50242.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain | SKP2 LOC100506406 NADK2 MIR580 UGT3A1 CAPSL SPEF2 IL7R UGT3A2 RANBP3L LMBRD2 |
nsv830262 | Chr.5:36074209 - 36293770 on Build GRCh38 | Gain | SKP2 NADK2 MIR580 RANBP3L LMBRD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |