Assay Details
Target Gene Details
Entrez Gene ID: | 7045 |
Gene Name: | transforming growth factor beta induced |
Gene Aliases: |
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1 |
Location: |
Chr.5:136028895-136063818 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGFBI | NM_000358.2 | NP_000349.1 | ||
AK093916.1 | ||||
AK094055.1 | ||||
AK094581.1 | ||||
AK222833.1 | BAD96553.1 | |||
AK298603.1 | ||||
AK303643.1 | ||||
BC000097.2 | AAH00097.1 | |||
BC004972.1 | AAH04972.1 | |||
BT009820.1 | AAP88822.1 | |||
DA051240.1 | ||||
KU178357.1 | ||||
KU178358.1 | ||||
M77349.1 | AAA61163.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759380 | Chr.5:135829912 - 136139310 on Build GRCh38 | Loss | SMAD5 IL9 VTRNA2-1 TGFBI LECT2 SLC25A48 FBXL21 SMAD5-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |