Assay Details
Target Gene Details
Entrez Gene ID: | 64374 |
Gene Name: | SIL1 nucleotide exchange factor |
Gene Aliases: |
BAP, MSS, ULG5 |
Location: |
Chr.5:138946720-139198376 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SIL1 | NM_001037633.1 | NP_001032722.1 | ||
NM_022464.4 | NP_071909.1 | |||
XM_011543570.2 | XP_011541872.1 | |||
AF547994.1 | AAN84477.1 | |||
AJ299442.1 | CAC17773.1 | |||
AK074624.1 | BAC11096.1 | |||
AK075177.1 | BAC11452.1 | |||
AY358950.1 | AAQ89309.1 | |||
BC011568.1 | AAH11568.1 | |||
DQ059315.1 | ||||
DQ173771.1 | ||||
DQ173772.1 | ||||
KU178743.1 | ||||
KU178744.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |