Assay Details
Target Gene Details
Entrez Gene ID: | 404734 |
Gene Name: | ANKHD1-EIF4EBP3 readthrough |
Gene Aliases: |
MASK-BP3, MASK-BP3ARF |
Location: |
Chr.5:140401814-140549578 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ANKHD1-EIF4EBP3 | NM_020690.5 | NP_065741.3 | ||
AB029008.1 | BAA83037.1 | |||
AF521883.1 | AAO14944.1 | |||
AK125734.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)