Assay Details
Target Gene Details
Entrez Gene ID: | 51128 |
Gene Name: | secretion associated Ras related GTPase 1B |
Gene Aliases: |
ANDD, CMRD, GTBPB, SARA2 |
Location: |
Chr.5:134601149-134632843 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAR1B | NM_001033503.2 | NP_001028675.1 | ||
NM_016103.3 | NP_057187.1 | |||
XM_017009521.1 | XP_016865010.1 | |||
AF087850.1 | AAP97161.1 | |||
AF092130.1 | AAD40372.1 | |||
AK056821.1 | ||||
AK223452.1 | BAD97172.1 | |||
AL512710.1 | CAC21652.1 | |||
BC002847.2 | AAH02847.1 | |||
BC093034.1 | AAH93034.1 | |||
CD048852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520381 | Chr.5:134523918 - 134643239 on Build GRCh38 | Gain+Loss |
![]() ![]() |
nsv830491 | Chr.5:134557253 - 134724800 on Build GRCh38 | Loss |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)