Assay Details
Target Gene Details
Entrez Gene ID: | 404203 |
Gene Name: | serine peptidase inhibitor, Kazal type 6 |
Gene Aliases: |
BUSI2, UNQ844 |
Location: |
Chr.5:148202794-148215137 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPINK6 | NM_001195290.1 | NP_001182219.1 | ||
NM_205841.3 | NP_995313.2 | |||
AY358716.1 | AAQ89078.1 | |||
BC032003.1 | AAH32003.1 | |||
BE122852.1 | ||||
GQ504704.1 | ||||
GQ504705.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3607101 | Chr.5:148020480 - 148213612 on Build GRCh38 | Loss | SPINK14 SPINK6 SPINK5 |
esv3607102 | Chr.5:148026584 - 148359140 on Build GRCh38 | Loss | LOC102546294 SPINK14 SPINK7 SPINK6 SPINK13 SPINK9 LOC105378220 SPINK5 |
nsv830515 | Chr.5:148081289 - 148222684 on Build GRCh38 | Gain+Loss | SPINK14 SPINK6 SPINK5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |