Assay Details
Target Gene Details
Entrez Gene ID: | 100505875 |
Gene Name: | long intergenic non-protein coding RNA 1088 |
Gene Aliases: |
- |
Location: |
Chr.4:78971748-79308799 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01088 | NR_038342.1 |
Target Gene Details
Entrez Gene ID: | 84779 |
Gene Name: | N(alpha)-acetyltransferase 11, NatA catalytic subunit |
Gene Aliases: |
ARD1B, ARD2, hARD2 |
Location: |
Chr.4:79155237-79326058 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA11 | XM_017008779.1 | XP_016864268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014428 | Chr.4:79168558 - 79211659 on Build GRCh38 | Gain | LINC01088 NAA11 |
esv2727859 | Chr.4:78716995 - 79557290 on Build GRCh38 | Deletion | LINC01088 NAA11 LOC101928893 MIR5096 BMP2K GK2 PAQR3 LINC00989 |
nsv1006026 | Chr.4:78111837 - 79966019 on Build GRCh38 | Loss | ANXA3 LOC101928893 PCAT4 GK2 LINC01088 NAA11 LINC01094 ANTXR2 MIR5096 BMP2K FRAS1 PAQR3 LINC00989 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |