Assay Details
Target Gene Details
Entrez Gene ID: | 7323 |
Gene Name: | ubiquitin conjugating enzyme E2 D3 |
Gene Aliases: |
E2(17)KB3, UBC4/5, UBCH5C |
Location: |
Chr.4:102794383-102868893 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2D3 | NM_001300795.1 | NP_001287724.1 | ||
NM_003340.6 | NP_003331.1 | |||
NM_181886.3 | NP_871615.1 | |||
NM_181887.2 | NP_871616.1 | |||
NM_181888.3 | NP_871617.1 | |||
NM_181889.2 | NP_871618.1 | |||
NM_181890.2 | NP_871619.1 | |||
NM_181891.2 | NP_871620.1 | |||
NM_181892.3 | NP_871621.1 | |||
NM_181893.2 | NP_871622.1 | |||
XM_017008584.1 | XP_016864073.1 | |||
AK095822.1 | BAC04632.1 | |||
AK127304.1 | ||||
AK315775.1 | ||||
AL833414.1 | ||||
AU131013.1 | ||||
BC003395.1 | AAH03395.1 | |||
BC037894.2 | AAH37894.1 | |||
BC066917.1 | AAH66917.1 | |||
BG613555.1 | ||||
BM541512.1 | ||||
BQ428985.1 | ||||
BU567649.1 | ||||
BU570661.1 | ||||
CR456916.1 | CAG33197.1 | |||
DA335600.1 | ||||
DA550074.1 | ||||
DA557036.1 | ||||
DA808523.1 | ||||
DA965437.1 | ||||
DB234695.1 | ||||
HY109217.1 | ||||
U39318.1 | AAA91461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526256 | Chr.4:102791432 - 102806853 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)