Assay Details
Target Gene Details
Entrez Gene ID: | 23284 |
Gene Name: | adhesion G protein-coupled receptor L3 |
Gene Aliases: |
CIRL3, CL3, LEC3, LPHN3 |
Location: |
Chr.4:61201256-62078335 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRL3 | NM_001322246.1 | NP_001309175.1 | ||
NM_001322402.1 | NP_001309331.1 | |||
NM_015236.5 | NP_056051.2 | |||
AK309793.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |