Assay Details
Target Gene Details
Entrez Gene ID: | 9407 |
Gene Name: | transmembrane protease, serine 11D |
Gene Aliases: |
HAT |
Location: |
Chr.4:67820876-67885063 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMPRSS11D | NM_004262.2 | NP_004253.1 | ||
XM_005265710.4 | XP_005265767.1 | |||
XM_017008851.1 | XP_016864340.1 | |||
AB002134.1 | BAA28691.1 | |||
AK094054.1 | ||||
AK290289.1 | ||||
AK296853.1 | ||||
BC125195.1 | ||||
BC125196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829960 | Chr.4:67783519 - 67943911 on Build GRCh38 | Loss | TMPRSS11D TMPRSS11A |
nsv1007376 | Chr.4:67827765 - 68142484 on Build GRCh38 | Gain | TMPRSS11F SYT14P1 LOC550113 TMPRSS11GP TMPRSS11D TMPRSS11A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |