Assay Details
Target Gene Details
Entrez Gene ID: | 246175 |
Gene Name: | CCR4-NOT transcription complex subunit 6 like |
Gene Aliases: |
CCR4b |
Location: |
Chr.4:77713387-77820238 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNOT6L | NM_001286790.1 | NP_001273719.1 | ||
NM_144571.2 | NP_653172.2 | |||
XM_011531806.2 | XP_011530108.1 | |||
XM_011531807.2 | XP_011530109.1 | |||
XM_011531808.2 | XP_011530110.1 | |||
XM_011531809.1 | XP_011530111.1 | |||
XM_011531810.1 | XP_011530112.1 | |||
XM_017007956.1 | XP_016863445.1 | |||
XM_017007957.1 | XP_016863446.1 | |||
AK027214.1 | ||||
AK303417.1 | ||||
AL133112.1 | CAB61415.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv518757 | Chr.4:77716111 - 77773984 on Build GRCh38 | Loss | CNOT6L |
nsv829976 | Chr.4:77655503 - 77836644 on Build GRCh38 | Gain | CNOT6L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |