Assay Details
Target Gene Details
Entrez Gene ID: | 22998 |
Gene Name: | LIM and calponin homology domains 1 |
Gene Aliases: |
LIMCH1A, LMO7B |
Location: |
Chr.4:41359607-41700044 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |