Assay Details
Target Gene Details
Entrez Gene ID: | 127 |
Gene Name: | alcohol dehydrogenase 4 (class II), pi polypeptide |
Gene Aliases: |
ADH-2, HEL-S-4 |
Location: |
Chr.4:99123657-99144330 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADH4 | NM_000670.4 | NP_000661.2 | ||
NM_001306171.1 | NP_001293100.1 | |||
NM_001306172.1 | NP_001293101.1 | |||
XM_017007713.1 | XP_016863202.1 | |||
XM_017007714.1 | XP_016863203.1 | |||
XM_017007715.1 | XP_016863204.1 | |||
AK290835.1 | ||||
AK295556.1 | ||||
AK313649.1 | ||||
BC022319.1 | AAH22319.1 | |||
CD014080.1 | ||||
EU668317.1 | ||||
M15943.1 | AAA51595.1 |
Target Gene Details
Entrez Gene ID: | 100507053 |
Gene Name: | uncharacterized LOC100507053 |
Gene Aliases: |
- |
Location: |
Chr.4:99088857-99301356 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100507053 | NR_037884.1 | |||
BX647987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1002927 | Chr.4:99031321 - 99227533 on Build GRCh38 | Gain | METAP1 ADH6 ADH4 PCNAP1 ADH5 LOC100507053 |
nsv1002574 | Chr.4:99102848 - 99212536 on Build GRCh38 | Loss | ADH6 ADH4 PCNAP1 LOC100507053 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |