Assay Details
Target Gene Details
Entrez Gene ID: | 9227 |
Gene Name: | lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase) |
Gene Aliases: |
LCA14 |
Location: |
Chr.4:154740841-154753119 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRAT | NM_001301645.1 | NP_001288574.1 | ||
BC031053.1 | AAH31053.1 | |||
DQ266889.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |