Assay Details
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001199021.1 | NP_001185950.1 | ||
DA201089.1 | ||||
DB249869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2726767 | Chr.4:1195098 - 1195251 on Build GRCh38 | Deletion | SPON2 |
dgv8891n54 | Chr.4:1181073 - 1261142 on Build GRCh38 | Loss | CTBP1 CTBP1-AS LOC100130872 LOC101928548 CTBP1-AS2 SPON2 |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | PDE6B CRIPAK SLC26A1 CTBP1-AS2 RNF212 TMEM175 LOC105374344 SPON2 MFSD7 ATP5I MAEA UVSSA LOC100130872 GAK LOC101928548 DGKQ CTBP1 FGFRL1 MYL5 CTBP1-AS IDUA TMED11P LOC107986211 LOC101928521 NKX1-1 CPLX1 LOC100129917 PCGF3 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | MAEA CTBP1 UVSSA CTBP1-AS CRIPAK LOC100130872 NKX1-1 LOC101928548 CTBP1-AS2 SPON2 |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | CTBP1 CTBP1-AS LOC100130872 LOC101928548 SPON2 |
nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion | MAEA CTBP1 UVSSA CTBP1-AS LOC100130872 LOC101928548 CTBP1-AS2 SPON2 |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | MAEA CTBP1 LOC105374344 CTBP1-AS TMED11P LOC100130872 LOC101928548 CTBP1-AS2 SPON2 RNF212 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |