Assay Details
Target Gene Details
Entrez Gene ID: | 11162 |
Gene Name: | nudix hydrolase 6 |
Gene Aliases: |
ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1 |
Location: |
Chr.4:122892644-122923004 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT6 | NM_007083.4 | NP_009014.2 | ||
NM_198041.2 | NP_932158.1 | |||
AB209758.1 | BAD92995.1 | |||
AF019632.2 | AAD01635.1 | |||
AF019633.2 | AAD01636.2 | |||
AK291871.1 | ||||
AK294450.1 | ||||
BC009842.2 | AAH09842.1 | |||
BG386740.1 | ||||
CB128685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894022 | Chr.4:122865924 - 123123191 on Build GRCh38 | Gain | NUDT6 SPATA5 FGF2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |