Assay Details
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001199021.1 | NP_001185950.1 | ||
DA087369.1 | ||||
DA201089.1 | ||||
DB249869.1 | ||||
DB254983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8891n54 | Chr.4:1181073 - 1261142 on Build GRCh38 | Loss | LOC101928548 CTBP1-AS2 CTBP1 CTBP1-AS SPON2 LOC100130872 |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | LOC100129917 IDUA CRIPAK MFSD7 LOC105374344 RNF212 NKX1-1 SLC26A1 LOC100130872 LOC101928548 PDE6B CTBP1 MAEA PCGF3 LOC101928521 CTBP1-AS MYL5 SPON2 UVSSA DGKQ CPLX1 CTBP1-AS2 LOC107986211 TMED11P ATP5I FGFRL1 GAK TMEM175 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | LOC101928548 CTBP1-AS2 CTBP1 MAEA NKX1-1 CTBP1-AS CRIPAK SPON2 UVSSA LOC100130872 |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | LOC101928548 CTBP1 CTBP1-AS SPON2 LOC100130872 |
nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion | LOC101928548 CTBP1-AS2 CTBP1 MAEA CTBP1-AS SPON2 UVSSA LOC100130872 |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | LOC101928548 CTBP1-AS2 CTBP1 TMED11P MAEA RNF212 CTBP1-AS SPON2 LOC105374344 LOC100130872 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |