Assay Details
Target Gene Details
Entrez Gene ID: | 10608 |
Gene Name: | MAX dimerization protein 4 |
Gene Aliases: |
MAD4, MST149, MSTP149, bHLHc12 |
Location: |
Chr.4:2247432-2262143 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MXD4 | NM_006454.2 | NP_006445.1 | ||
XM_011513388.2 | XP_011511690.1 | |||
AF040963.1 | AAB97009.1 | |||
AF082740.1 | AAF98553.1 | |||
AF187724.1 | AAQ13695.1 | |||
AK024501.1 | 1 | 1680 | BAB15791.1 | |
BC002713.2 | AAH02713.1 | |||
BC068060.1 | AAH68060.1 | |||
BT019969.1 | AAV38772.1 | |||
BT019970.1 | AAV38773.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508990 | Chr.4:2246239 - 2324327 on Build GRCh38 | Insertion | MIR4800 ZFYVE28 MXD4 |
nsv593377 | Chr.4:2246106 - 2261919 on Build GRCh38 | Loss | MIR4800 MXD4 |
nsv4200 | Chr.4:2252081 - 2286377 on Build GRCh38 | Insertion | ZFYVE28 MXD4 |
nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain | MIR4800 POLN RNF4 ZFYVE28 CFAP99 HAUS3 MXD4 |
esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion | NOP14 NAT8L MIR4800 C4orf48 POLN TNIP2 ADD1 MFSD10 NOP14-AS1 SH3BP2 RNF4 ZFYVE28 CFAP99 HAUS3 MXD4 FAM193A |
nsv527189 | Chr.4:2243138 - 2276689 on Build GRCh38 | Loss | MIR4800 ZFYVE28 MXD4 |
dgv8910n54 | Chr.4:2249015 - 2262496 on Build GRCh38 | Loss | MIR4800 MXD4 |
nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss | MIR4800 POLN ZFYVE28 HAUS3 MXD4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |