Assay Details
Target Gene Details
Entrez Gene ID: | 100130872 |
Gene Name: | uncharacterized LOC100130872 |
Gene Aliases: |
- |
Location: |
Chr.4:1195783-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100130872 | NR_024569.1 | |||
CR627052.1 |
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001199021.1 | NP_001185950.1 | ||
DA201089.1 | ||||
DB249869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv8891n54 | Chr.4:1181073 - 1261142 on Build GRCh38 | Loss | LOC100130872 SPON2 CTBP1-AS2 LOC101928548 CTBP1 CTBP1-AS |
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | GAK LOC100129917 LOC107986211 TMEM175 LOC101928521 NKX1-1 PCGF3 MYL5 LOC105374344 LOC100130872 SPON2 CTBP1-AS2 PDE6B LOC101928548 CRIPAK CPLX1 DGKQ MFSD7 MAEA CTBP1 UVSSA IDUA SLC26A1 RNF212 FGFRL1 TMED11P CTBP1-AS ATP5I |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | LOC100130872 SPON2 CTBP1-AS2 NKX1-1 LOC101928548 CRIPAK CTBP1 UVSSA CTBP1-AS MAEA |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | LOC100130872 SPON2 LOC101928548 CTBP1 CTBP1-AS |
nsv508988 | Chr.4:1190955 - 1358870 on Build GRCh38 | Insertion | LOC100130872 SPON2 CTBP1-AS2 LOC101928548 CTBP1 UVSSA CTBP1-AS MAEA |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | LOC100130872 SPON2 CTBP1-AS2 LOC101928548 CTBP1 RNF212 TMED11P CTBP1-AS MAEA LOC105374344 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |