Assay Details
Target Gene Details
Entrez Gene ID: | 419 |
Gene Name: | ADP-ribosyltransferase 3 |
Gene Aliases: |
ARTC3 |
Location: |
Chr.4:76011180-76112802 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ART3 | NM_001130017.2 | NP_001123489.1 | ||
AK225860.1 | ||||
AY543656.1 | AAT10278.1 | |||
AY543657.1 | AAT10279.1 | |||
AY543658.1 | AAT10280.1 | |||
AY543659.1 | AAT10281.1 | |||
DB451386.1 | ||||
U47054.1 | AAB01894.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |