Assay Details
Target Gene Details
Entrez Gene ID: | 3355 |
Gene Name: | 5-hydroxytryptamine receptor 1F |
Gene Aliases: |
5-HT-1F, 5-HT1F, 5HT6, HTR1EL, MR77 |
Location: |
Chr.3:87792767-87994856 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTR1F | NM_001322208.1 | NP_001309137.1 | ||
NM_001322209.1 | NP_001309138.1 | |||
NM_001322210.1 | NP_001309139.1 | |||
XM_011533664.2 | XP_011531966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv527279 | Chr.3:86796784 - 87814130 on Build GRCh38 | Gain | MIR4795 VGLL3 POU1F1 LINC00506 HTR1F CHMP2B |
nsv590918 | Chr.3:87274184 - 87861781 on Build GRCh38 | Gain | POU1F1 HTR1F |
nsv834756 | Chr.3:87714643 - 87889029 on Build GRCh38 | Loss | HTR1F |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |