Assay Details
Target Gene Details
Entrez Gene ID: | 1371 |
Gene Name: | coproporphyrinogen oxidase |
Gene Aliases: |
CPO, CPX, HCP |
Location: |
Chr.3:98568448-98593684 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CPOX |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834775 | Chr.3:98451215 - 98648708 on Build GRCh38 | Gain | OR5K1 CLDND1 CPOX OR5K2 GPR15 |
nsv591065 | Chr.3:98485354 - 99047482 on Build GRCh38 | Gain | ST3GAL6 CLDND1 DCBLD2 ST3GAL6-AS1 CPOX OR5K2 GPR15 |
dgv293n21 | Chr.3:98382257 - 98598992 on Build GRCh38 | Loss | OR5K1 CLDND1 OR5K3 CPOX OR5K2 GPR15 |
dgv4819n100 | Chr.3:98565439 - 98662629 on Build GRCh38 | Gain | CPOX |