Assay Details
Target Gene Details
Entrez Gene ID: | 905 |
Gene Name: | cyclin T2 |
Gene Aliases: |
CYCT2 |
Location: |
Chr.2:134918810-134959345 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCNT2 | NM_001241.3 | NP_001232.1 | ||
NM_001320748.1 | NP_001307677.1 | |||
NM_001320749.1 | NP_001307678.1 | |||
NM_058241.2 | NP_490595.1 | |||
NR_037649.1 | ||||
NR_135468.1 | ||||
XM_017005227.1 | XP_016860716.1 | |||
XM_017005228.1 | XP_016860717.1 | |||
XM_017005229.1 | XP_016860718.1 | |||
XM_017005230.1 | XP_016860719.1 | |||
XM_017005231.1 | XP_016860720.1 | |||
XM_017005232.1 | XP_016860721.1 | |||
AF048731.1 | AAC39665.1 | |||
AF048732.1 | AAC39666.1 | |||
AK292913.1 | ||||
AK294889.1 | ||||
BC114366.2 | AAI14367.1 | |||
BI769278.1 | ||||
BX648174.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |