Assay Details
Target Gene Details
Entrez Gene ID: | 9173 |
Gene Name: | interleukin 1 receptor like 1 |
Gene Aliases: |
DER4, FIT-1, IL33R, ST2, ST2L, ST2V, T1 |
Location: |
Chr.2:102311502-102352367 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IL1RL1 | NM_001282408.1 | NP_001269337.1 | ||
NM_016232.4 | NP_057316.3 | |||
XM_011512151.1 | XP_011510453.1 | |||
AB012701.3 | ||||
AK303389.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2673990 | Chr.2:102314830 - 102320561 on Build GRCh38 | Deletion | IL1RL1 |
esv3591787 | Chr.2:102314919 - 102320486 on Build GRCh38 | Loss | IL1RL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |