Assay Details
Target Gene Details
Entrez Gene ID: | 2571 |
Gene Name: | glutamate decarboxylase 1 |
Gene Aliases: |
CPSQ1, GAD, SCP |
Location: |
Chr.2:170813210-170861151 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAD1 | XM_011510922.1 | XP_011509224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528929 | Chr.2:170815681 - 170821869 on Build GRCh38 | Loss | GAD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |