Assay Details
Target Gene Details
Entrez Gene ID: | 51719 |
Gene Name: | calcium binding protein 39 |
Gene Aliases: |
CGI-66, MO25 |
Location: |
Chr.2:230712842-230821075 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CAB39 | NM_001130849.1 | NP_001124321.1 | ||
NM_001130850.1 | NP_001124322.1 | |||
NM_016289.3 | NP_057373.1 | |||
XM_011511350.2 | XP_011509652.1 | |||
AK292382.1 | ||||
AK315692.1 | ||||
BC020570.1 | AAH20570.1 | |||
BG773506.1 | ||||
DB455871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv584675 | Chr.2:230699911 - 230801231 on Build GRCh38 | Loss | LOC151475 CAB39 |
nsv834565 | Chr.2:230697957 - 230920661 on Build GRCh38 | Gain | LOC151475 GPR55 ITM2C LOC151484 CAB39 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |