Assay Details
Target Gene Details
Entrez Gene ID: | 51540 |
Gene Name: | selenocysteine lyase |
Gene Aliases: |
SCL, hSCL |
Location: |
Chr.2:238060924-238099413 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SCLY | NM_016510.5 | NP_057594.4 | ||
AB209458.2 | BAD92695.1 | |||
AF175767.1 | AAF36816.1 | |||
AK293283.1 | ||||
AK298409.1 | ||||
AK301009.1 | ||||
BC000586.2 | AAH00586.2 | |||
BC007891.2 | AAH07891.1 | |||
DN995828.1 |
Target Gene Details
Entrez Gene ID: | 100533179 |
Gene Name: | UBE2F-SCLY readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.2:237966945-238099413 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBE2F-SCLY | NR_037904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss | ASB1 MLPH TRAF3IP1 LOC105373958 ERFE RAMP1 ACKR3 SCLY HES6 IQCA1 COL6A3 ASB18 LRRFIP1 KLHL30 UBE2F COPS8 LOC151174 LINC01107 MIR6811 LOC105373944 RBM44 LOC105373991 LOC643387 AGAP1 PER2 PRLH LOC93463 RAB17 LOC101927958 ESPNL GBX2 UBE2F-SCLY ILKAP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |