Assay Details
Target Gene Details
Entrez Gene ID: | 115361 |
Gene Name: | guanylate binding protein 4 |
Gene Aliases: |
Mpa2 |
Location: |
Chr.1:89181148-89198950 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GBP4 | NM_052941.4 | NP_443173.2 | ||
AF288814.1 | AAL02054.1 | |||
AK131094.1 | ||||
AK304245.1 | ||||
AK312417.1 | ||||
AL832576.2 | CAD89936.1 | |||
BC017889.1 | ||||
BC070055.1 | AAH70055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830548 | Chr.1:89142066 - 89298597 on Build GRCh38 | Gain | LOC729930 GBP5 GBP4 GBP7 |
nsv546796 | Chr.1:89052437 - 89256179 on Build GRCh38 | Loss | GBP2 GBP4 GBP7 GBP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |