Assay Details
Target Gene Details
Entrez Gene ID: | 56833 |
Gene Name: | SLAM family member 8 |
Gene Aliases: |
BLAME, CD353, SBBI42 |
Location: |
Chr.1:159826650-159837497 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLAMF8 | NM_020125.2 | NP_064510.1 | ||
XM_005245345.2 | XP_005245402.1 | |||
XM_011509775.1 | XP_011508077.1 | |||
XM_011509776.2 | XP_011508078.1 | |||
XM_011509777.2 | XP_011508079.1 | |||
XM_011509778.2 | XP_011508080.1 | |||
XM_011509779.2 | XP_011508081.1 | |||
XM_011509780.2 | XP_011508082.2 | |||
XM_017001844.1 | XP_016857333.1 | |||
XM_017001845.1 | XP_016857334.1 | |||
XM_017001846.1 | XP_016857335.1 | |||
AF146761.1 | AAF67470.1 | |||
AK074669.1 | BAC11123.1 | |||
BC109194.2 | AAI09195.1 | |||
DA578369.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain | SLAMF8 MIR4259 VSIG8 C1orf204 FCRL6 CRP OR10J5 CFAP45 TAGLN2 APCS DUSP23 IGSF9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |