Assay Details
Target Gene Details
Entrez Gene ID: | 4914 |
Gene Name: | neurotrophic receptor tyrosine kinase 1 |
Gene Aliases: |
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA |
Location: |
Chr.1:156815750-156881850 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NTRK1 | NM_001007792.1 | NP_001007793.1 | ||
AK126428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss | RRNAD1 CRABP2 HDGF MRPL24 ISG20L2 PRCC SH2D2A INSRR PEAR1 NTRK1 |
nsv831659 | Chr.1:156771159 - 156980204 on Build GRCh38 | Loss | ARHGEF11 MIR765 LRRC71 PRCC SH2D2A INSRR PEAR1 NTRK1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |