Assay Details
Target Gene Details
Entrez Gene ID: | 84871 |
Gene Name: | ATP/GTP binding protein like 4 |
Gene Aliases: |
CCP6 |
Location: |
Chr.1:48520157-50024011 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGBL4 | XM_017002595.1 | XP_016858084.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585972 | Chr.1:48526674 - 48530109 on Build GRCh38 | Loss | AGBL4 |
esv3585970 | Chr.1:48459872 - 48603418 on Build GRCh38 | Loss | SPATA6 AGBL4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |