Assay Details
Target Gene Details
Entrez Gene ID: | 2052 |
Gene Name: | epoxide hydrolase 1 |
Gene Aliases: |
EPHX, EPOX, HYL1, MEH |
Location: |
Chr.1:225810074-225845563 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHX1 | NM_001136018.3 | NP_001129490.1 | ||
NM_001291163.1 | NP_001278092.1 | |||
AK313436.1 | ||||
BC003567.1 | AAH03567.1 | |||
BC008291.1 | AAH08291.1 | |||
BC095430.1 | AAH95430.1 | |||
BU541640.1 | ||||
HY180507.1 | ||||
KF032815.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524126 | Chr.1:225786075 - 226022952 on Build GRCh38 | Gain | MIR6741 LEFTY2 PYCR2 LEFTY1 TMEM63A SRP9 EPHX1 SDE2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |