Assay Details
Target Gene Details
Entrez Gene ID: | 57549 |
Gene Name: | immunoglobulin superfamily member 9 |
Gene Aliases: |
FP18798, IGSF9A, Nrt1 |
Location: |
Chr.1:159927039-159945661 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGSF9 | NM_001135050.1 | NP_001128522.1 | ||
NM_020789.3 | NP_065840.2 | |||
XM_017001884.1 | XP_016857373.1 | |||
AB037776.1 | ||||
AY358128.1 | AAQ88495.1 | |||
BC030141.2 | AAH30141.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524006 | Chr.1:159916632 - 159953321 on Build GRCh38 | Gain | IGSF9 SLAMF9 TAGLN2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |