Assay Details
Target Gene Details
Entrez Gene ID: | 64801 |
Gene Name: | ARV1 homolog, fatty acid homeostasis modulator |
Gene Aliases: |
- |
Location: |
Chr.1:230979075-231000733 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARV1 | NM_022786.1 | NP_073623.1 | ||
XM_017002106.1 | XP_016857595.1 | |||
AF271780.1 | AAG44791.1 | |||
AF290878.1 | AAG47671.1 | |||
AF321442.1 | AAK11180.1 | |||
AK293049.1 | ||||
BC016309.1 | AAH16309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1011066 | Chr.1:230940432 - 231242850 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)