Assay Details
Target Gene Details
Entrez Gene ID: | 84871 |
Gene Name: | ATP/GTP binding protein like 4 |
Gene Aliases: |
CCP6 |
Location: |
Chr.1:48520157-50024011 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGBL4 | NM_001323573.1 | NP_001310502.1 | ||
NM_001323574.1 | NP_001310503.1 | |||
NM_001323575.1 | NP_001310504.1 | |||
NM_032785.3 | NP_116174.3 | |||
NR_136623.1 | ||||
XM_011542308.2 | XP_011540610.1 | |||
XM_017002595.1 | XP_016858084.1 | |||
XM_017002596.1 | XP_016858085.1 | |||
XM_017002597.1 | XP_016858086.1 | |||
AK094826.1 | ||||
AK294394.1 | ||||
AK294633.1 | ||||
AK296221.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585970 | Chr.1:48459872 - 48603418 on Build GRCh38 | Loss | AGBL4 SPATA6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |