Assay Details
Target Gene Details
Entrez Gene ID: | 23421 |
Gene Name: | integrin subunit beta 3 binding protein |
Gene Aliases: |
CENP-R, CENPR, HSU37139, NRIF3, TAP20 |
Location: |
Chr.1:63440770-63523517 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ITGB3BP | NM_001206739.1 | NP_001193668.1 | ||
NM_014288.4 | NP_055103.3 | |||
NR_045147.1 | ||||
XM_006710514.3 | XP_006710577.3 | |||
XM_006710515.3 | XP_006710578.3 | |||
XM_017000853.1 | XP_016856342.1 | |||
XM_017000854.1 | XP_016856343.1 | |||
XM_017000855.1 | XP_016856344.1 | |||
XM_017000857.1 | XP_016856346.1 | |||
AF175306.1 | AAF00239.1 | |||
AK309622.1 | ||||
AK312944.1 | ||||
BC009929.2 | AAH09929.1 | |||
BC014385.1 | AAH14385.1 | |||
BU853146.1 | ||||
U37139.1 | AAC50294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv546434 | Chr.1:63449672 - 63576116 on Build GRCh38 | Loss | ITGB3BP EFCAB7 DLEU2L |
nsv470714 | Chr.1:63301926 - 63537185 on Build GRCh38 | Gain | LINC00466 FOXD3-AS1 ITGB3BP EFCAB7 ALG6 MIR6068 FOXD3 |
nsv546433 | Chr.1:63288065 - 63537185 on Build GRCh38 | Gain | LINC00466 FOXD3-AS1 ITGB3BP EFCAB7 ALG6 MIR6068 FOXD3 |
nsv1002554 | Chr.1:63354605 - 63459148 on Build GRCh38 | Gain | ITGB3BP ALG6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |