Assay Details
Target Gene Details
Entrez Gene ID: | 8613 |
Gene Name: | phospholipid phosphatase 3 |
Gene Aliases: |
Dri42, LPP3, PAP2B, PPAP2B, VCIP |
Location: |
Chr.1:56494747-56579584 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLPP3 | NM_003713.4 | NP_003704.3 | ||
AB000889.1 | BAA22594.1 | |||
AF017786.1 | AAC63433.1 | |||
AF043329.1 | AAD02271.1 | |||
AF480883.1 | AAO84481.1 | |||
AK123105.1 | ||||
AK312439.1 | ||||
BC009196.2 | AAH09196.1 | |||
BT019589.1 | AAV38396.1 | |||
U79294.1 | AAB50222.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |