Assay Details
Target Gene Details
Entrez Gene ID: | 1952 |
Gene Name: | cadherin EGF LAG seven-pass G-type receptor 2 |
Gene Aliases: |
ADGRC2, CDHF10, EGFL2, Flamingo1, MEGF3 |
Location: |
Chr.1:109249705-109275756 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CELSR2 | NM_001408.2 | NP_001399.1 | ||
XM_005270580.4 | XP_005270637.1 | |||
AF234887.1 | AAG00080.1 | |||
D87469.2 | BAA13407.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv993086 | Chr.1:109254248 - 109263222 on Build GRCh38 | Deletion | CELSR2 |
nsv517354 | Chr.1:109239415 - 109283514 on Build GRCh38 | Loss | PSRC1 CELSR2 |
nsv1012267 | Chr.1:109168205 - 109279887 on Build GRCh38 | Gain | SARS PSRC1 KIAA1324 CELSR2 |
nsv951381 | Chr.1:109248679 - 109277078 on Build GRCh38 | Deletion | CELSR2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |