Assay Details
Target Gene Details
Entrez Gene ID: | 5733 |
Gene Name: | prostaglandin E receptor 3 |
Gene Aliases: |
EP3, EP3-I, EP3-II, EP3-III, EP3-IV, EP3-VI, EP3e, PGE2-R |
Location: |
Chr.1:70852353-71047814 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGER3 | NM_198714.1 | NP_942007.1 | ||
NM_198716.1 | NP_942009.1 | |||
NM_198717.1 | NP_942010.1 | |||
NR_028292.1 | ||||
NR_028293.1 | ||||
NR_028294.1 | ||||
XM_011541810.2 | XP_011540112.1 | |||
AK290634.1 | ||||
AY387856.1 | AAQ90021.1 | |||
AY429108.1 | AAR07903.1 | |||
BC024229.1 | AAH24229.1 | |||
BC118578.1 | ||||
BC118659.1 | ||||
D38298.1 | BAA07417.1 | |||
D38300.1 | BAA07419.1 | |||
D38301.1 | BAA07420.1 | |||
D86097.1 | BAA19958.1 | |||
D86098.1 | BAA19959.1 | |||
L27489.1 | AAC13373.1 | |||
S69326.1 | AAB30208.1 | |||
U13214.1 | AAA21130.1 | |||
U13215.1 | AAA21131.1 | |||
U13216.1 | AAA21132.1 | |||
X83857.1 | CAA58737.1 | |||
X83859.1 | CAA58739.1 | |||
X83861.1 | CAA58741.1 | |||
X83863.1 | CAA58743.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv22e214 | Chr.1:70885560 - 70901337 on Build GRCh38 | Loss | PTGER3 |
nsv830148 | Chr.1:70844683 - 71018249 on Build GRCh38 | Gain | PTGER3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |