Assay Details
Target Gene Details
Entrez Gene ID: | 1969 |
Gene Name: | EPH receptor A2 |
Gene Aliases: |
ARCC2, CTPA, CTPP1, CTRCT6, ECK |
Location: |
Chr.1:16124337-16156104 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHA2 | XM_017000534.1 | XP_016856023.1 | ||
XM_017000535.1 | XP_016856024.1 | |||
XM_017000536.1 | XP_016856025.1 | |||
AK296788.1 | ||||
BC008655.1 | AAH08655.1 | |||
BC037166.2 | AAH37166.1 | |||
M59371.1 | AAA53375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833869 | Chr.1:16110258 - 16278634 on Build GRCh38 | Loss | FBXO42 EPHA2 ANO7P1 ARHGEF19 RSG1 |
nsv520148 | Chr.1:16005414 - 16139508 on Build GRCh38 | Loss | FAM131C HSPB7 C1orf64 EPHA2 CLCNKA CLCNKB |
nsv508936 | Chr.1:15915852 - 16181025 on Build GRCh38 | Insertion | FAM131C HSPB7 C1orf64 EPHA2 CLCNKA CLCNKB SPEN ZBTB17 |
nsv833758 | Chr.1:15969028 - 16140519 on Build GRCh38 | Gain+Loss | FAM131C HSPB7 C1orf64 EPHA2 CLCNKA CLCNKB ZBTB17 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |