Assay Details
Target Gene Details
Entrez Gene ID: | 114784 |
Gene Name: | CUB and Sushi multiple domains 2 |
Gene Aliases: |
dJ1007G16.1, dJ1007G16.2, dJ947L8.1 |
Location: |
Chr.1:33513999-34165842 on Build GRCh38 |
Assay Gene Location: | Within Intron 71 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CSMD2 | NM_001281956.1 | NP_001268885.1 | ||
NM_052896.4 | NP_443128.2 | |||
XM_017000185.1 | XP_016855674.1 | |||
XM_017000186.1 | XP_016855675.1 | |||
XM_017000187.1 | XP_016855676.1 | |||
XM_017000188.1 | XP_016855677.1 | |||
XM_017000190.1 | XP_016855679.1 | |||
XM_017000194.1 | XP_016855683.1 | |||
AB067471.1 | BAB67777.1 | |||
AB212622.1 | BAD97692.1 | |||
AK095627.1 | BAC04593.1 | |||
AY210418.1 | AAO34701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3585687 | Chr.1:33404909 - 33620947 on Build GRCh38 | Gain | ZSCAN20 PHC2 CSMD2 |
esv3585686 | Chr.1:33404224 - 33959487 on Build GRCh38 | Gain | CSMD2-AS1 ZSCAN20 PHC2 HMGB4 CSMD2 |