Assay Details
Target Gene Details
Entrez Gene ID: | 92344 |
Gene Name: | golgin, RAB6 interacting |
Gene Aliases: |
GO, NTKLBP1, SCYL1BP1 |
Location: |
Chr.1:170532122-170553834 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GORAB | NM_001146039.1 | 4 | 1082 | NP_001139511.1 |
NM_001320252.1 | NP_001307181.1 | |||
NM_152281.2 | NP_689494.2 | |||
NR_027397.1 | ||||
XM_006711628.3 | XP_006711691.1 | |||
XM_011510149.2 | XP_011508451.1 | |||
XM_011510150.2 | XP_011508452.1 | |||
XM_011510151.1 | XP_011508453.1 | |||
XM_017002807.1 | XP_016858296.1 | |||
AF143889.1 | AAD32702.1 | |||
AK021814.1 | BAB13903.1 | |||
AK057661.1 | ||||
AK308339.1 | ||||
BC047476.1 | AAH47476.1 | |||
BC064945.1 | 4 | 1073 | AAH64945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3476 | Chr.1:170534143 - 170567828 on Build GRCh38 | Insertion | GORAB |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |