Assay Details
Target Gene Details
Entrez Gene ID: | 5321 |
Gene Name: | phospholipase A2 group IVA |
Gene Aliases: |
PLA2G4, cPLA2, cPLA2-alpha |
Location: |
Chr.1:186828900-186989896 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G4A | NM_001311193.1 | NP_001298122.1 | ||
NM_024420.2 | NP_077734.1 | |||
XM_011509642.2 | XP_011507944.1 | |||
AK290336.1 | ||||
AK302938.1 | ||||
BC114340.2 | AAI14341.1 | |||
DA451491.1 | ||||
M68874.1 | AAA60105.1 | |||
M72393.1 | AAB00789.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2762854 | Chr.1:186577969 - 186878442 on Build GRCh38 | Gain | PTGS2 PLA2G4A PACERR |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |