Assay Details
Target Gene Details
Entrez Gene ID: | 55160 |
Gene Name: | Rho guanine nucleotide exchange factor 10 like |
Gene Aliases: |
GrinchGEF |
Location: |
Chr.1:17520197-17697875 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGEF10L | NM_018125.3 | NP_060595.3 | ||
NR_137287.1 | ||||
NR_137288.1 | ||||
XM_005245923.2 | XP_005245980.2 | |||
XM_005245925.2 | XP_005245982.2 | |||
XM_006710728.1 | XP_006710791.1 | |||
XM_006710729.1 | XP_006710792.1 | |||
XM_006710731.1 | XP_006710794.1 | |||
XM_017001617.1 | XP_016857106.1 | |||
AB046846.1 | ||||
AK129959.1 | ||||
AK311076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834302 | Chr.1:17531435 - 17713452 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_next.png)
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)