Assay Details
Target Gene Details
Entrez Gene ID: | 85397 |
Gene Name: | regulator of G-protein signaling 8 |
Gene Aliases: |
- |
Location: |
Chr.1:182641814-182753291 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RGS8 | NM_001102450.2 | NP_001095920.1 | ||
NM_033345.3 | NP_203131.1 | |||
XM_005245555.3 | XP_005245612.1 | |||
XM_011510089.2 | XP_011508391.1 | |||
XM_011510090.2 | XP_011508392.1 | |||
XM_011510091.2 | XP_011508393.1 | |||
XM_017002631.1 | XP_016858120.1 | |||
XM_017002632.1 | XP_016858121.1 | |||
XM_017002633.1 | XP_016858122.1 | |||
XM_017002634.1 | XP_016858123.1 | |||
XM_017002635.1 | XP_016858124.1 | |||
XM_017002636.1 | XP_016858125.1 | |||
XM_017002637.1 | XP_016858126.1 | |||
AF300649.1 | AAG18443.1 | |||
AK294660.1 | ||||
AK296514.1 | ||||
BC069677.1 | AAH69677.1 | |||
BC069718.1 | AAH69718.1 | |||
BC103865.1 | AAI03866.1 | |||
BC103866.1 | AAI03867.1 | |||
BC103867.1 | AAI03868.1 | |||
BX281622.1 | ||||
BX954018.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3810 | Chr.1:182630610 - 182661590 on Build GRCh38 | Deletion | RGS8 |
nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain | LINC01344 NPL SHCBP1L LAMC1 RGS8 LOC647070 LOC284648 RGS16 ZNF648 LINC00272 RNASEL GLUL TEDDM1 LOC105371644 RGSL1 DHX9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |