Assay Details
Target Gene Details
Entrez Gene ID: | 3765 |
Gene Name: | potassium voltage-gated channel subfamily J member 9 |
Gene Aliases: |
GIRK3, KIR3.3 |
Location: |
Chr.1:160055001-160090563 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KCNJ9 | XM_017001247.1 | XP_016856736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520189 | Chr.1:160041722 - 160125835 on Build GRCh38 | Loss | KCNJ10 KCNJ9 ATP1A2 IGSF8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |