Assay Details
Target Gene Details
Entrez Gene ID: | 5792 |
Gene Name: | protein tyrosine phosphatase, receptor type F |
Gene Aliases: |
BNAH2, LAR |
Location: |
Chr.1:43522238-43623672 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv157n100 | Chr.1:43525952 - 43606749 on Build GRCh38 | Gain | PTPRF |
nsv519570 | Chr.1:43539609 - 43606395 on Build GRCh38 | Loss | PTPRF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |