Assay Details
Target Gene Details
Entrez Gene ID: | 4921 |
Gene Name: | discoidin domain receptor tyrosine kinase 2 |
Gene Aliases: |
MIG20a, NTRKR3, TKT, TYRO10 |
Location: |
Chr.1:162631213-162783524 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDR2 | NM_001014796.1 | NP_001014796.1 | ||
NM_006182.2 | NP_006173.2 | |||
XM_006711344.3 | XP_006711407.1 | |||
XM_011509586.2 | XP_011507888.1 | |||
XM_011509587.2 | XP_011507889.1 | |||
XM_011509588.2 | XP_011507890.1 | |||
AK095975.1 | ||||
AK225063.1 | ||||
AK225196.1 | ||||
AK314388.1 | ||||
BC052998.2 | AAH52998.1 | |||
BX640899.1 | CAE45946.1 | |||
CF994640.1 | ||||
X74764.1 | CAA52777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv516482 | Chr.1:162138329 - 162905405 on Build GRCh38 | Gain+Loss | CCDC190 HSD17B7 C1orf226 UAP1 UHMK1 SH2D1B NOS1AP MIR556 DDR2 C1orf111 MIR4654 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |